The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017019531.1 | XP_016875020.1 | 2 - 3 | 607 | 129 |
GenBank mRNA | AK001160.1 | - | 2 - 3 | 308 | 129 |