The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170695.1 | NP_001164166.1 | 3 - 4 | 350 | 129 |
NM_001170697.1 | NP_001164168.1 | 3 - 4 | 356 | 129 | |
NM_001170698.1 | NP_001164169.1 | 3 - 4 | 371 | 129 | |
NM_001170699.1 | NP_001164170.1 | 3 - 4 | 590 | 129 | |
NM_001321337.1 | NP_001308266.1 | 3 - 4 | 688 | 129 | |
NM_032598.4 | NP_115987.2 | 3 - 4 | 412 | 129 | |
GenBank mRNA | AF367472.1 | - | 3 - 4 | 304 | 129 |
AK301892.1 | - | 3 - 4 | 556 | 129 | |
AY035867.1 | - | 3 - 4 | 353 | 129 | |
AY035868.1 | - | 3 - 4 | 412 | 129 | |
BC029483.1 | - | 3 - 4 | 297 | 129 | |
BG716998.1 | - | 3 - 4 | 354 | 129 | |
BI463957.1 | - | 3 - 4 | 376 | 129 | |
BP371122.1 | - | 3 - 4 | 356 | 129 |