Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- VENTXP7
- Entrez Gene ID
-
391518
- Gene Name
- VENT homeobox pseudogene 7
- Gene Aliases
-
HPX42, VENTX1
- Chromosome Location
-
Chr.3: 21405726 - 21406708 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002311.1 |
|
1 - 1 |
555 |
131 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101927829 |
LOC101927829 |
|
-1.1mb
|
Sense |
Chr. - 20342468 - 20350928 on Build GRCh38 |
Downstream |
zinc finger protein 385D |
ZNF385D |
ZNF659 |
5.5kb
|
Anti-sense |
Chr. - 21418419 - 22372641 on Build GRCh38 |
View Full Product Details