The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126340.2 | NP_001119812.1 | 2 - 3 | 248 | 65 |
NM_001271818.1 | NP_001258747.1 | 2 - 3 | 271 | 65 | |
GenBank mRNA | AK056497.1 | - | 2 - 3 | 203 | 65 |
BC069270.1 | - | 1 - 2 | 42 | 65 | |
CN366874.1 | - | 2 - 3 | 167 | 65 | |
DA770788.1 | - | 2 - 3 | 244 | 65 | |
HY064081.1 | - | 2 - 3 | 249 | 65 |