The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270766.1 | NP_001257695.1 | 5 - 6 | 489 | 118 |
NM_001270767.1 | NP_001257696.1 | 4 - 5 | 423 | 118 | |
NM_173474.3 | NP_775745.1 | 6 - 7 | 592 | 118 | |
XM_011522355.2 | XP_011520657.1 | 3 - 4 | 427 | 118 | |
GenBank mRNA | AF092440.1 | - | 6 - 7 | 499 | 118 |
BC017336.2 | - | 6 - 7 | 489 | 118 | |
DC383993.1 | - | 5 - 6 | 437 | 118 |