Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ETV4
Entrez Gene ID
2118
Gene Name
ETS variant 4
Gene Aliases
E1A-F, E1AF, PEA3, PEAS3
Chromosome Location
Chr.17: 43527843 - 43546432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079675.2 NP_001073143.1 3 - 4 452 123
NM_001261437.1 NP_001248366.1 2 - 3 341 123
NM_001261438.1 NP_001248367.1 2 - 3 170 123
NM_001986.2 NP_001977.1 3 - 4 281 123
XM_011524514.1 XP_011522816.1 1 - 2 93 123
XM_017024349.1 XP_016879838.1 3 - 4 243 123
GenBank mRNA AK290429.1 - 3 - 4 243 123
AK298872.1 - 2 - 3 249 123
AK299019.1 - 2 - 3 132 123
AK312934.1 - 3 - 4 243 123
AK315961.1 - 2 - 3 232 123
BC016623.1 - 3 - 4 223 123
BM013535.1 - 3 - 4 281 123
BP268896.1 - 3 - 4 359 123
DC375462.1 - 3 - 4 453 123
DQ396625.1 - 1 - 2 160 123
DQ396626.1 - 1 - 2 126 123
DR156106.1 - 3 - 4 449 123
EU693079.1 - - 353 123
U18018.1 - 3 - 4 356 123

View Full Product Details