Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUK
Entrez Gene ID
197258
Gene Name
fucokinase
Gene Aliases
1110046B12Rik
Chromosome Location
Chr.16: 70454421 - 70480274 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145059.2 NP_659496.2 22 - 23 2991 99
XM_006721162.1 XP_006721225.1 21 - 22 2921 99
XM_011522928.1 XP_011521230.1 22 - 23 3169 99
XM_011522929.1 XP_011521231.1 22 - 23 3034 99
XM_017023012.1 XP_016878501.1 22 - 23 3018 99
XM_017023013.1 XP_016878502.1 19 - 20 2814 99
XM_017023014.1 XP_016878503.1 17 - 18 2946 99
GenBank mRNA AJ441184.1 - 22 - 23 2990 99
AK056456.1 - 22 - 23 2990 99
AK092025.1 - 14 - 15 2140 99
AK096727.1 - 14 - 15 2197 99
AK128387.1 - 22 - 23 3009 99
AK315590.1 - 22 - 23 2994 99
BC013735.1 - 20 - 21 2777 99
BC032542.1 - 22 - 23 2997 99

View Full Product Details