The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000285.3 | NP_000276.2 | 13 - 14 | 1285 | 74 |
NM_001166056.1 | NP_001159528.1 | 11 - 12 | 1162 | 74 | |
NM_001166057.1 | NP_001159529.1 | 11 - 12 | 1093 | 74 | |
GenBank mRNA | AK290756.1 | - | 13 - 14 | 1166 | 74 |
AK290781.1 | - | 13 - 14 | 1181 | 74 | |
AK291561.1 | - | 13 - 14 | 1183 | 74 | |
AK293126.1 | - | 11 - 12 | 988 | 74 | |
AK294619.1 | - | 11 - 12 | 1060 | 74 | |
BC004305.1 | - | 13 - 14 | 1156 | 74 | |
BC015027.1 | - | 13 - 14 | 1186 | 74 | |
BC028295.1 | - | 13 - 14 | 1157 | 74 | |
BT006692.1 | - | 13 - 14 | 1152 | 74 | |
CR541669.1 | - | 13 - 14 | 1152 | 74 | |
J04605.1 | - | 13 - 14 | 1168 | 74 |