Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164750.1 NP_001158222.1 23 - 24 2067 91
NM_004318.3 NP_004309.2 23 - 24 2262 91
XM_005251235.2 XP_005251292.1 24 - 25 2276 91
XM_005251236.2 XP_005251293.1 24 - 25 2273 91
XM_005251238.2 XP_005251295.1 23 - 24 2231 91
XM_005251239.2 XP_005251296.1 23 - 24 2219 91
XM_005251240.1 XP_005251297.1 24 - 25 2054 91
XM_005251242.2 XP_005251299.1 22 - 23 2174 91
XM_005251243.2 XP_005251300.1 23 - 24 2147 91
XM_005251244.1 XP_005251301.1 23 - 24 1994 91
XM_005251246.2 XP_005251303.1 22 - 23 2102 91
XM_005251247.2 XP_005251304.1 22 - 23 2090 91
XM_005251248.1 XP_005251305.1 22 - 23 1860 91
XM_005251250.2 XP_005251307.1 21 - 22 2045 91
XM_017013419.1 XP_016868908.1 24 - 25 2276 91
XM_017013420.1 XP_016868909.1 24 - 25 2273 91
XM_017013421.1 XP_016868910.1 23 - 24 2234 91
XM_017013422.1 XP_016868911.1 23 - 24 2231 91
XM_017013423.1 XP_016868912.1 23 - 24 2231 91
XM_017013424.1 XP_016868913.1 23 - 24 2228 91
XM_017013425.1 XP_016868914.1 23 - 24 2219 91
XM_017013426.1 XP_016868915.1 24 - 25 2054 91
XM_017013427.1 XP_016868916.1 24 - 25 2051 91
XM_017013428.1 XP_016868917.1 22 - 23 2174 91
XM_017013429.1 XP_016868918.1 23 - 24 2228 91
XM_017013430.1 XP_016868919.1 23 - 24 2147 91
XM_017013431.1 XP_016868920.1 23 - 24 1917 91
XM_017013432.1 XP_016868921.1 23 - 24 2144 91
XM_017013433.1 XP_016868922.1 23 - 24 1914 91
XM_017013434.1 XP_016868923.1 23 - 24 2216 91
XM_017013435.1 XP_016868924.1 23 - 24 1994 91
XM_017013436.1 XP_016868925.1 22 - 23 2189 91
XM_017013437.1 XP_016868926.1 22 - 23 2102 91
XM_017013438.1 XP_016868927.1 23 - 24 2144 91
XM_017013439.1 XP_016868928.1 23 - 24 1914 91
XM_017013440.1 XP_016868929.1 23 - 24 1925 91
XM_017013441.1 XP_016868930.1 22 - 23 2099 91
XM_017013442.1 XP_016868931.1 22 - 23 2087 91
XM_017013443.1 XP_016868932.1 21 - 22 2042 91
XM_017013444.1 XP_016868933.1 22 - 23 1788 91
GenBank mRNA AK304314.1 - 23 - 24 2023 91
S83325.1 - 23 - 24 2004 91
U03109.1 - 23 - 24 2067 91

View Full Product Details