Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164750.1 NP_001158222.1 8 - 9 779 154
NM_001164753.1 NP_001158225.1 8 - 9 695 154
NM_001164755.1 NP_001158227.1 7 - 8 845 154
NM_004318.3 NP_004309.2 8 - 9 974 154
NM_032466.3 NP_115855.1 8 - 9 974 154
NM_032468.4 NP_115857.1 9 - 10 824 154
XM_005251235.2 XP_005251292.1 9 - 10 988 154
XM_005251238.2 XP_005251295.1 9 - 10 988 154
XM_005251240.1 XP_005251297.1 9 - 10 766 154
XM_005251243.2 XP_005251300.1 8 - 9 859 154
XM_005251246.2 XP_005251303.1 7 - 8 814 154
XM_017013419.1 XP_016868908.1 9 - 10 988 154
XM_017013422.1 XP_016868911.1 8 - 9 943 154
XM_017013423.1 XP_016868912.1 9 - 10 988 154
XM_017013426.1 XP_016868915.1 9 - 10 766 154
XM_017013430.1 XP_016868919.1 8 - 9 859 154
XM_017013431.1 XP_016868920.1 8 - 9 629 154
XM_017013437.1 XP_016868926.1 7 - 8 814 154
XM_017013440.1 XP_016868929.1 8 - 9 637 154
XM_017013444.1 XP_016868933.1 7 - 8 500 154
XM_017013445.1 XP_016868934.1 8 - 9 629 154
XM_017013447.1 XP_016868936.1 7 - 8 500 154
GenBank mRNA AF289489.1 - 8 - 9 716 154
AF306765.1 - 9 - 10 745 154
AK291836.1 - 8 - 9 787 154
AK295528.1 - 8 - 9 659 154
AK304314.1 - 8 - 9 735 154
BC025236.1 - 7 - 8 773 154
EU718494.1 - 8 - 9 782 154
S83325.1 - 8 - 9 716 154
U03109.1 - 8 - 9 782 154

View Full Product Details