Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ASPH
- Entrez Gene ID
-
444
- Gene Name
- aspartate beta-hydroxylase
- Gene Aliases
-
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
- Chromosome Location
-
Chr.8: 61500556 - 61714640 on Build GRCh38
View Full Product Details