Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC5A7
Entrez Gene ID
60482
Gene Name
solute carrier family 5 member 7
Gene Aliases
CHT, CHT1, HMN7A
Chromosome Location
Chr.2: 107986514 - 108014497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305005.2 NP_001291934.1 2 - 3 475 86
NM_001305007.2 NP_001291936.1 2 - 3 478 86
NM_021815.4 NP_068587.1 2 - 3 478 86
GenBank mRNA AB043997.1 - 2 - 3 453 86
AF276871.1 - 1 - 2 177 86
AJ401466.1 - 1 - 2 195 86
AK315280.1 - 2 - 3 467 86
BC111524.1 - 2 - 3 420 86
BC111525.1 - 2 - 3 425 86
DA714970.1 - 2 - 3 479 86

View Full Product Details