Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC5A7
Entrez Gene ID
60482
Gene Name
solute carrier family 5 member 7
Gene Aliases
CHT, CHT1, HMN7A
Chromosome Location
Chr.2: 107986514 - 108014497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305005.2 NP_001291934.1 6 - 7 1039 86
NM_001305006.2 NP_001291935.1 5 - 6 813 86
NM_001305007.2 NP_001291936.1 6 - 7 1005 86
NM_021815.4 NP_068587.1 6 - 7 1042 86
XM_011511580.2 XP_011509882.1 4 - 5 593 86
XM_017004628.1 XP_016860117.1 4 - 5 1813 86
XM_017004629.1 XP_016860118.1 4 - 5 4321 86
GenBank mRNA AB043997.1 - 6 - 7 1017 86
AF276871.1 - 5 - 6 741 86
AJ401466.1 - 5 - 6 759 86
AK300801.1 - 5 - 6 803 86
AK315280.1 - 6 - 7 1031 86
BC111524.1 - 6 - 7 947 86
BC111525.1 - 6 - 7 989 86

View Full Product Details