The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000459.4 | NP_000450.2 | 19 - 20 | 3514 | 97 |
NM_001290077.1 | NP_001277006.1 | 18 - 19 | 3385 | 97 | |
NM_001290078.1 | NP_001277007.1 | 17 - 18 | 3070 | 97 | |
XM_005251561.1 | XP_005251618.1 | 19 - 20 | 3178 | 97 | |
XM_005251563.1 | XP_005251620.1 | 18 - 19 | 3049 | 97 | |
GenBank mRNA | AB208796.1 | - | 19 - 20 | 3192 | 97 |
AK291775.1 | - | 19 - 20 | 3202 | 97 | |
AK294887.1 | - | 17 - 18 | 3071 | 97 | |
AK295043.1 | - | 18 - 19 | 3111 | 97 | |
BC035514.1 | - | 19 - 20 | 3189 | 97 | |
L06139.1 | - | 19 - 20 | 3212 | 97 |