The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007215.3 | NP_009146.2 | 4 - 5 | 1067 | 101 |
XM_017024072.1 | XP_016879561.1 | 4 - 5 | 1020 | 101 | |
GenBank mRNA | AF142992.1 | - | 4 - 5 | 1050 | 101 |
AF177201.1 | - | 4 - 5 | 969 | 101 | |
AF184344.1 | - | 4 - 5 | 1050 | 101 | |
AK307801.1 | - | 4 - 5 | 1020 | 101 | |
BC000913.2 | - | 4 - 5 | 815 | 101 | |
BC009194.2 | - | 4 - 5 | 1037 | 101 |