The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195098.1 | NP_001182027.1 | 16 - 17 | 1786 | 114 |
NM_001195099.1 | NP_001182028.1 | 14 - 15 | 1477 | 114 | |
NM_015199.3 | NP_056014.2 | 16 - 17 | 1742 | 114 | |
XM_005264996.3 | XP_005265053.1 | 16 - 17 | 3113 | 114 | |
XM_011533540.2 | XP_011531842.1 | 16 - 17 | 3122 | 114 | |
XM_011533541.2 | XP_011531843.1 | 16 - 17 | 3122 | 114 | |
XM_011533542.2 | XP_011531844.1 | 15 - 16 | 1926 | 114 | |
XM_011533543.2 | XP_011531845.1 | 15 - 16 | 2921 | 114 | |
XM_011533545.2 | XP_011531847.1 | 16 - 17 | 2449 | 114 | |
XM_011533546.2 | XP_011531848.1 | 16 - 17 | 2440 | 114 | |
XM_011533547.2 | XP_011531849.1 | 15 - 16 | 2398 | 114 | |
XM_017006026.1 | XP_016861515.1 | 17 - 18 | 2520 | 114 | |
XM_017006027.1 | XP_016861516.1 | 16 - 17 | 2469 | 114 | |
XM_017006028.1 | XP_016861517.1 | 16 - 17 | 3122 | 114 | |
XM_017006029.1 | XP_016861518.1 | 15 - 16 | 1527 | 114 | |
GenBank mRNA | AB002377.2 | - | 16 - 17 | 1618 | 114 |
AK126888.1 | - | 16 - 17 | 2004 | 114 | |
AK293770.1 | - | 16 - 17 | 1787 | 114 | |
AK295813.1 | - | 15 - 16 | 1612 | 114 | |
AY367056.1 | - | 14 - 15 | 1476 | 114 | |
BC106948.2 | - | 16 - 17 | 1816 | 114 | |
BC113868.1 | - | 16 - 17 | 1805 | 114 | |
BC114476.1 | - | 16 - 17 | 1816 | 114 | |
BQ423818.1 | - | 2 - 3 | 158 | 114 |