Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
Chromosome Location
Chr.3: 15667236 - 15859814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195098.1 NP_001182027.1 7 - 8 880 149
NM_001195099.1 NP_001182028.1 5 - 6 571 149
NM_015199.3 NP_056014.2 7 - 8 836 149
XM_005264996.3 XP_005265053.1 7 - 8 2207 149
XM_011533540.2 XP_011531842.1 7 - 8 2216 149
XM_011533541.2 XP_011531843.1 7 - 8 2216 149
XM_011533542.2 XP_011531844.1 6 - 7 1020 149
XM_011533543.2 XP_011531845.1 6 - 7 2015 149
XM_011533545.2 XP_011531847.1 7 - 8 1543 149
XM_011533546.2 XP_011531848.1 7 - 8 1534 149
XM_011533547.2 XP_011531849.1 6 - 7 1492 149
XM_017006026.1 XP_016861515.1 8 - 9 1614 149
XM_017006027.1 XP_016861516.1 7 - 8 1563 149
XM_017006028.1 XP_016861517.1 7 - 8 2216 149
XM_017006029.1 XP_016861518.1 6 - 7 614 149
GenBank mRNA AB002377.2 - 7 - 8 712 149
AK293770.1 - 7 - 8 881 149
AK295813.1 - 6 - 7 706 149
AY367056.1 - 5 - 6 571 149
BC106948.2 - 7 - 8 910 149
BC113868.1 - 7 - 8 899 149
BC114476.1 - 7 - 8 910 149

View Full Product Details