Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
Chromosome Location
Chr.3: 15667236 - 15859814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195098.1 NP_001182027.1 8 - 9 1084 98
NM_001195099.1 NP_001182028.1 6 - 7 775 98
NM_015199.3 NP_056014.2 8 - 9 1040 98
XM_005264996.3 XP_005265053.1 8 - 9 2411 98
XM_011533540.2 XP_011531842.1 8 - 9 2420 98
XM_011533541.2 XP_011531843.1 8 - 9 2420 98
XM_011533542.2 XP_011531844.1 7 - 8 1224 98
XM_011533543.2 XP_011531845.1 7 - 8 2219 98
XM_011533545.2 XP_011531847.1 8 - 9 1747 98
XM_011533546.2 XP_011531848.1 8 - 9 1738 98
XM_011533547.2 XP_011531849.1 7 - 8 1696 98
XM_017006026.1 XP_016861515.1 9 - 10 1818 98
XM_017006027.1 XP_016861516.1 8 - 9 1767 98
XM_017006028.1 XP_016861517.1 8 - 9 2420 98
XM_017006029.1 XP_016861518.1 7 - 8 818 98
GenBank mRNA AB002377.2 - 8 - 9 916 98
AK126888.1 - 8 - 9 1302 98
AK293770.1 - 8 - 9 1085 98
AK295813.1 - 7 - 8 910 98
AY367056.1 - 6 - 7 775 98
BC106948.2 - 8 - 9 1114 98
BC113868.1 - 8 - 9 1103 98
BC114476.1 - 8 - 9 1114 98

View Full Product Details