The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020397.3 | NP_065130.1 | 1 - 2 | 371 | 90 |
NM_153498.3 | NP_705718.1 | 1 - 2 | 371 | 90 | |
XM_006717482.3 | XP_006717545.1 | 1 - 2 | 400 | 90 | |
XM_006717483.3 | XP_006717546.1 | 1 - 2 | 362 | 90 | |
GenBank mRNA | AB081726.1 | - | 1 - 2 | 327 | 90 |
AF286366.1 | - | 1 - 2 | 182 | 90 | |
BC035745.1 | - | 1 - 2 | 188 | 90 | |
BU542765.1 | - | 1 - 2 | 432 | 90 | |
HY168807.1 | - | 1 - 2 | 372 | 90 |