The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020397.3 | NP_065130.1 | 3 - 4 | 576 | 75 |
NM_153498.3 | NP_705718.1 | 3 - 4 | 576 | 75 | |
XM_006717481.3 | XP_006717544.1 | 3 - 4 | 840 | 75 | |
XM_006717482.3 | XP_006717545.1 | 3 - 4 | 605 | 75 | |
XM_006717483.3 | XP_006717546.1 | 3 - 4 | 567 | 75 | |
XM_011519591.2 | XP_011517893.1 | 3 - 4 | 1013 | 75 | |
XM_011519592.2 | XP_011517894.1 | 2 - 3 | 1205 | 75 | |
XM_011519593.2 | XP_011517895.1 | 2 - 3 | 1253 | 75 | |
XM_011519595.2 | XP_011517897.1 | 2 - 3 | 1067 | 75 | |
XM_017016438.1 | XP_016871927.1 | 4 - 5 | 871 | 75 | |
XM_017016439.1 | XP_016871928.1 | 5 - 6 | 702 | 75 | |
GenBank mRNA | AB081726.1 | - | 3 - 4 | 532 | 75 |
AF286366.1 | - | 3 - 4 | 387 | 75 | |
AK289393.1 | - | 4 - 5 | 387 | 75 | |
BC035745.1 | - | 3 - 4 | 393 | 75 | |
BU542765.1 | - | 3 - 4 | 637 | 75 |