The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000700.2 | NP_000691.1 | 10 - 11 | 1012 | 109 |
XM_011518609.1 | XP_011516911.1 | 10 - 11 | 939 | 109 | |
XM_017014657.1 | XP_016870146.1 | 10 - 11 | 958 | 109 | |
GenBank mRNA | AB451274.1 | - | 9 - 10 | 804 | 109 |
AB451401.1 | - | 9 - 10 | 804 | 109 | |
AK074480.1 | - | 9 - 10 | 1686 | 109 | |
BC001275.1 | - | 10 - 11 | 856 | 109 | |
BC035993.1 | - | 10 - 11 | 878 | 109 | |
BT019896.1 | - | 9 - 10 | 804 | 109 | |
BT019916.1 | - | 9 - 10 | 804 | 109 | |
BT019917.1 | - | 9 - 10 | 804 | 109 | |
CR407684.1 | - | 9 - 10 | 804 | 109 | |
X05908.1 | - | 10 - 11 | 878 | 109 |