Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
Chromosome Location
Chr.9: 73151731 - 73170393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000700.2 NP_000691.1 12 - 13 1194 137
XM_011518609.1 XP_011516911.1 12 - 13 1121 137
XM_017014657.1 XP_016870146.1 12 - 13 1140 137
GenBank mRNA AB451274.1 - 11 - 12 986 137
AB451401.1 - 11 - 12 986 137
AK074480.1 - 11 - 12 1868 137
BC001275.1 - 12 - 13 1038 137
BC034157.1 - 1 - 2 277 137
BC035993.1 - 12 - 13 1060 137
BT019896.1 - 11 - 12 986 137
BT019916.1 - 11 - 12 986 137
BT019917.1 - 11 - 12 986 137
CR407684.1 - 11 - 12 986 137
X05908.1 - 12 - 13 1060 137

View Full Product Details