Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
Chromosome Location
Chr.9: 73151731 - 73170393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000700.2 NP_000691.1 7 - 8 765 67
XM_011518609.1 XP_011516911.1 7 - 8 692 67
XM_017014657.1 XP_016870146.1 7 - 8 711 67
GenBank mRNA AB451274.1 - 6 - 7 557 67
AB451401.1 - 6 - 7 557 67
AK074480.1 - 6 - 7 1439 67
AK296808.1 - 7 - 8 680 67
BC001275.1 - 7 - 8 609 67
BC035993.1 - 7 - 8 631 67
BT019896.1 - 6 - 7 557 67
BT019916.1 - 6 - 7 557 67
BT019917.1 - 6 - 7 557 67
CR407684.1 - 6 - 7 557 67
X05908.1 - 7 - 8 631 67

View Full Product Details