Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A11
Entrez Gene ID
55867
Gene Name
solute carrier family 22 member 11
Gene Aliases
OAT4, hOAT4
Chromosome Location
Chr.11: 64555601 - 64572875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307985.1 NP_001294914.1 2 - 3 892 82
NM_018484.3 NP_060954.1 2 - 3 892 82
GenBank mRNA AB026116.1 - 2 - 3 545 82
AK075224.1 - 2 - 3 238 82
AK129930.1 - 2 - 3 804 82
AK222869.1 - 2 - 3 550 82
AK290791.1 - 2 - 3 875 82
AK309931.1 - 2 - 3 552 82
BC034384.1 - 2 - 3 867 82

View Full Product Details