The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005257410.3 | XP_005257467.1 | 1 - 2 | 85 | 93 |
XM_005257411.3 | XP_005257468.1 | 1 - 2 | 85 | 93 | |
XM_005257412.3 | XP_005257469.1 | 1 - 2 | 85 | 93 | |
XM_005257415.3 | XP_005257472.1 | 1 - 2 | 85 | 93 | |
GenBank mRNA | BX647976.1 | - | 1 - 2 | 93 | 93 |