The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004661.3 | NP_004652.2 | 11 - 12 | 1315 | 98 |
GenBank mRNA | AB011472.1 | - | 11 - 12 | 1302 | 98 |
AF053977.1 | - | 11 - 12 | 1282 | 98 | |
AF191341.1 | - | 11 - 12 | 1266 | 98 | |
AK223250.1 | - | 11 - 12 | 1303 | 98 | |
AK291610.1 | - | 11 - 12 | 1297 | 98 | |
AK304635.1 | - | 10 - 11 | 1179 | 98 | |
BC017713.1 | - | 11 - 12 | 1274 | 98 | |
BT009810.1 | - | 11 - 12 | 1266 | 98 |