The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258426.1 | NP_001245355.1 | 1 - 2 | 264 | 131 |
NM_001258427.1 | NP_001245356.1 | 1 - 2 | 264 | 131 | |
NM_031467.2 | NP_113655.2 | 1 - 2 | 264 | 131 | |
XM_005268521.3 | XP_005268578.2 | 1 - 2 | 821 | 131 | |
XM_017009936.1 | XP_016865425.1 | 1 - 2 | 821 | 131 | |
XM_017009939.1 | XP_016865428.1 | 1 - 2 | 821 | 131 | |
XM_017009943.1 | XP_016865432.1 | 1 - 2 | 318 | 131 | |
GenBank mRNA | AB032762.1 | - | 1 - 2 | 223 | 131 |
AF336237.1 | - | 1 - 2 | 256 | 131 | |
BC136262.1 | - | 1 - 2 | 255 | 131 | |
BC143602.1 | - | 1 - 2 | 255 | 131 |