The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109891.1 | NP_001103361.1 | 7 - 8 | 1142 | 94 |
NM_002746.2 | NP_002737.2 | 8 - 9 | 1274 | 94 | |
GenBank mRNA | AK091009.1 | - | 7 - 8 | 1966 | 94 |
AK307995.1 | - | 8 - 9 | 1188 | 94 | |
BC000205.1 | - | 3 - 4 | 394 | 94 | |
BC013992.1 | - | 8 - 9 | 1185 | 94 | |
BX537897.1 | - | 7 - 8 | 1965 | 94 | |
M84490.1 | - | 8 - 9 | 1135 | 94 | |
X60188.1 | - | 9 - 10 | 1246 | 94 | |
Z11696.1 | - | 8 - 9 | 1102 | 94 |