The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000689.4 | NP_000680.2 | 7 - 8 | 1067 | 62 |
GenBank mRNA | AB209821.1 | - | 7 - 8 | 793 | 62 |
AF003341.1 | - | 7 - 8 | 750 | 62 | |
AK026641.1 | - | 7 - 8 | 794 | 62 | |
AK293176.1 | - | 7 - 8 | 760 | 62 | |
AK302168.1 | - | 6 - 7 | 578 | 62 | |
AY390731.1 | - | 7 - 8 | 750 | 62 | |
BC001505.2 | - | 7 - 8 | 775 | 62 | |
BT006921.1 | - | 7 - 8 | 750 | 62 | |
EU794589.1 | - | 7 - 8 | 750 | 62 | |
FJ224286.1 | - | 7 - 8 | 803 | 62 | |
K03000.1 | - | 3 - 4 | 267 | 62 | |
M26761.1 | - | 3 - 4 | 267 | 62 |