Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNA2
Entrez Gene ID
5362
Gene Name
plexin A2
Gene Aliases
OCT, PLXN2
Chromosome Location
Chr.1: 208022242 - 208244384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025179.3 NP_079455.3 17 - 18 4012 74
XM_005273164.2 XP_005273221.1 17 - 18 4076 74
XM_005273165.3 XP_005273222.1 17 - 18 4076 74
GenBank mRNA AB007932.1 - 17 - 18 3463 74
BC132676.1 - 17 - 18 3367 74
BC136530.1 - 17 - 18 3367 74
X87831.2 - 15 - 16 1917 74

View Full Product Details