Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNA2
Entrez Gene ID
5362
Gene Name
plexin A2
Gene Aliases
OCT, PLXN2
Chromosome Location
Chr.1: 208022242 - 208244384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025179.3 NP_079455.3 21 - 22 4775 67
XM_005273164.2 XP_005273221.1 21 - 22 4839 67
XM_005273165.3 XP_005273222.1 21 - 22 4839 67
GenBank mRNA AB007932.1 - 21 - 22 4226 67
BC009343.2 - 4 - 5 765 67
BC132676.1 - 21 - 22 4130 67
BC136530.1 - 21 - 22 4130 67
X87831.2 - 19 - 20 2680 67

View Full Product Details