Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
Chromosome Location
Chr.11: 417930 - 442011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012302.2 NP_001012302.2 11 - 12 1003 66
XM_011520048.1 XP_011518350.1 11 - 12 1003 66
XM_011520051.2 XP_011518353.1 11 - 12 1003 66
XM_011520052.2 XP_011518354.1 11 - 12 1003 66
XM_017017644.1 XP_016873133.1 11 - 12 1003 66
XM_017017645.1 XP_016873134.1 11 - 12 1003 66
XM_017017646.1 XP_016873135.1 11 - 12 1003 66
XM_017017647.1 XP_016873136.1 11 - 12 1003 66
GenBank mRNA AK026923.1 - 11 - 12 950 66
AK074088.1 - 6 - 7 2785 66
AL832289.1 - 1 - 2 192 66
BC128557.1 - 11 - 12 971 66
BC144415.1 - 10 - 11 1029 66

View Full Product Details