The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012302.2 | NP_001012302.2 | 13 - 14 | 1267 | 83 |
XM_011520048.1 | XP_011518350.1 | 13 - 14 | 1267 | 83 | |
XM_011520051.2 | XP_011518353.1 | 13 - 14 | 1267 | 83 | |
XM_011520053.2 | XP_011518355.1 | 13 - 14 | 1363 | 83 | |
GenBank mRNA | AK026923.1 | - | 13 - 14 | 83 | |
AK074088.1 | - | 8 - 9 | 83 | ||
AK096874.1 | - | 4 - 5 | 83 | ||
AL832289.1 | - | 3 - 4 | 83 | ||
BC128557.1 | - | 13 - 14 | 83 | ||
BC144415.1 | - | 12 - 13 | 83 |