Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
Chromosome Location
Chr.11: 417930 - 442011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012302.2 NP_001012302.2 14 - 15 1308 71
XM_011520048.1 XP_011518350.1 14 - 15 1308 71
XM_011520051.2 XP_011518353.1 14 - 15 1308 71
XM_011520053.2 XP_011518355.1 14 - 15 1404 71
GenBank mRNA AK026923.1 - 14 - 15 1255 71
AK074088.1 - 9 - 10 3090 71
AK096874.1 - 5 - 6 596 71
AL832289.1 - 4 - 5 497 71
BC128557.1 - 14 - 15 1276 71
BC144415.1 - 13 - 14 1334 71

View Full Product Details