The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000726.3 | NP_000717.2 | 12 - 13 | 1184 | 78 |
NM_001005746.2 | NP_001005746.1 | 12 - 13 | 1134 | 78 | |
NM_001005747.2 | NP_001005747.1 | 11 - 12 | 1272 | 78 | |
NM_001320722.1 | NP_001307651.1 | 12 - 13 | 1432 | 78 | |
XM_011511795.2 | XP_011510097.1 | 12 - 13 | 2901 | 78 | |
XM_011511796.2 | XP_011510098.1 | 11 - 12 | 1855 | 78 | |
XM_011511797.2 | XP_011510099.1 | 9 - 10 | 1904 | 78 | |
XM_017004880.1 | XP_016860369.1 | 9 - 10 | 996 | 78 | |
XM_017004881.1 | XP_016860370.1 | 8 - 9 | 1064 | 78 | |
XM_017004882.1 | XP_016860371.1 | 9 - 10 | 2389 | 78 | |
XM_017004883.1 | XP_016860372.1 | 13 - 14 | 2723 | 78 | |
XM_017004884.1 | XP_016860373.1 | 11 - 12 | 2557 | 78 | |
XM_017004885.1 | XP_016860374.1 | 10 - 11 | 2397 | 78 | |
XM_017004886.1 | XP_016860375.1 | 3 - 4 | 477 | 78 | |
GenBank mRNA | AF038852.1 | - | 12 - 13 | 1094 | 78 |
AK290049.1 | - | 11 - 12 | 1253 | 78 | |
AK291378.1 | - | 13 - 14 | 1267 | 78 | |
AK294398.1 | - | 12 - 13 | 1108 | 78 | |
AK316045.1 | - | 12 - 13 | 1125 | 78 | |
AY054985.1 | - | 11 - 12 | 1243 | 78 | |
BC075049.2 | - | 12 - 13 | 1135 | 78 | |
U95020.1 | - | 12 - 13 | 1184 | 78 |