Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAD51
Entrez Gene ID
5888
Gene Name
RAD51 recombinase
Gene Aliases
BRCC5, FANCR, HRAD51, HsRad51, HsT16930, MRMV2, RAD51A, RECA
Chromosome Location
Chr.15: 40694774 - 40732340 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164270.1 NP_001157742.1 4 - 5 642 134
NM_002875.4 NP_002866.2 4 - 5 642 134
XM_006720626.3 XP_006720689.1 4 - 5 693 134
XM_011521857.2 XP_011520159.1 4 - 5 392 134
XM_011521858.2 XP_011520160.1 4 - 5 439 134
XM_011521859.2 XP_011520161.1 4 - 5 576 134
XM_011521860.2 XP_011520162.1 4 - 5 487 134
XM_011521861.2 XP_011520163.1 4 - 5 392 134
GenBank mRNA AK313503.1 - 4 - 5 597 134
AY425955.1 - 3 - 4 310 134
BT019705.1 - 3 - 4 343 134
CR536559.1 - 3 - 4 343 134
D13804.1 - 4 - 5 554 134
D14134.1 - 4 - 5 575 134
EU362635.1 - 4 - 5 642 134

View Full Product Details