Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr.X: 53532096 - 53686729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031407.6 NP_113584.3 36 - 37 4865 103
XM_005261965.3 XP_005262022.1 35 - 36 4932 103
XM_011530747.1 XP_011529049.1 37 - 38 5108 103
XM_011530751.2 XP_011529053.1 36 - 37 4976 103
XM_017029191.1 XP_016884680.1 35 - 36 7860 103
XM_017029192.1 XP_016884681.1 35 - 36 7860 103
XM_017029193.1 XP_016884682.1 35 - 36 7839 103
XM_017029194.1 XP_016884683.1 35 - 36 7860 103
XM_017029195.1 XP_016884684.1 35 - 36 7860 103
XM_017029196.1 XP_016884685.1 35 - 36 7860 103
XM_017029197.1 XP_016884686.1 35 - 36 7860 103
XM_017029198.1 XP_016884687.1 36 - 37 5202 103
XM_017029199.1 XP_016884688.1 37 - 38 5129 103
XM_017029200.1 XP_016884689.1 36 - 37 4999 103
XM_017029201.1 XP_016884690.1 36 - 37 5016 103
XM_017029202.1 XP_016884691.1 36 - 37 4992 103
XM_017029203.1 XP_016884692.1 36 - 37 4998 103
XM_017029204.1 XP_016884693.1 34 - 35 7610 103
XM_017029205.1 XP_016884694.1 34 - 35 7590 103
XM_017029206.1 XP_016884695.1 35 - 36 7860 103
GenBank mRNA AB002310.3 - 5 - 6 967 103
AB071605.1 - 9 - 10 1565 103
AK056671.1 - 19 - 20 3091 103
AK091352.1 - 8 - 9 1383 103
AK294575.1 - 23 - 24 3465 103
AY772009.1 - 34 - 35 4510 103
AY929612.1 - 33 - 34 4462 103
DQ097177.1 - 36 - 37 4864 103

View Full Product Details