The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031407.6 | NP_113584.3 | 82 - 83 | 13233 | 66 |
XM_005261965.3 | XP_005262022.1 | 81 - 82 | 13300 | 66 | |
XM_011530747.1 | XP_011529049.1 | 83 - 84 | 13476 | 66 | |
XM_011530751.2 | XP_011529053.1 | 82 - 83 | 13344 | 66 | |
XM_017029191.1 | XP_016884680.1 | 81 - 82 | 16228 | 66 | |
XM_017029192.1 | XP_016884681.1 | 81 - 82 | 16225 | 66 | |
XM_017029193.1 | XP_016884682.1 | 81 - 82 | 16207 | 66 | |
XM_017029194.1 | XP_016884683.1 | 81 - 82 | 16183 | 66 | |
XM_017029195.1 | XP_016884684.1 | 81 - 82 | 16180 | 66 | |
XM_017029196.1 | XP_016884685.1 | 81 - 82 | 16177 | 66 | |
XM_017029197.1 | XP_016884686.1 | 80 - 81 | 16129 | 66 | |
XM_017029198.1 | XP_016884687.1 | 82 - 83 | 13570 | 66 | |
XM_017029199.1 | XP_016884688.1 | 83 - 84 | 13497 | 66 | |
XM_017029200.1 | XP_016884689.1 | 82 - 83 | 13367 | 66 | |
XM_017029201.1 | XP_016884690.1 | 82 - 83 | 13384 | 66 | |
XM_017029202.1 | XP_016884691.1 | 82 - 83 | 13360 | 66 | |
XM_017029203.1 | XP_016884692.1 | 82 - 83 | 13366 | 66 | |
XM_017029204.1 | XP_016884693.1 | 80 - 81 | 15978 | 66 | |
XM_017029205.1 | XP_016884694.1 | 79 - 80 | 15856 | 66 | |
XM_017029206.1 | XP_016884695.1 | 78 - 79 | 15826 | 66 | |
GenBank mRNA | AB002310.3 | - | 51 - 52 | 9284 | 66 |
AB071605.1 | - | 55 - 56 | 9933 | 66 | |
AF057569.1 | - | 5 - 6 | 747 | 66 | |
AF161390.1 | - | 28 - 29 | 5480 | 66 | |
AK223167.1 | - | 7 - 8 | 1025 | 66 | |
AY772009.1 | - | 80 - 81 | 12878 | 66 | |
AY929612.1 | - | 79 - 80 | 12830 | 66 | |
BC002602.2 | - | 7 - 8 | 1034 | 66 | |
BC063505.1 | - | 6 - 7 | 873 | 66 | |
BC072421.1 | - | 11 - 12 | 1548 | 66 | |
BC107576.1 | - | 4 - 5 | 705 | 66 | |
CR456813.1 | - | 4 - 5 | 632 | 66 | |
DQ097177.1 | - | 82 - 83 | 13232 | 66 |