Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HUWE1
- Entrez Gene ID
-
10075
- Gene Name
- HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
- Gene Aliases
-
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
- Chromosome Location
-
Chr.X: 53532096 - 53686729 on Build GRCh38
View Full Product Details