The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142448.1 | NP_001135920.1 | 7 - 8 | 947 | 69 |
NM_001142450.1 | NP_001135922.1 | 7 - 8 | 1097 | 69 | |
NM_032038.2 | NP_114427.1 | 6 - 7 | 1184 | 69 | |
XM_017023757.1 | XP_016879246.1 | 7 - 8 | 970 | 69 | |
XM_017023758.1 | XP_016879247.1 | 7 - 8 | 966 | 69 | |
XM_017023759.1 | XP_016879248.1 | 7 - 8 | 950 | 69 | |
GenBank mRNA | AF212371.1 | - | 6 - 7 | 1131 | 69 |
AF370423.1 | - | 7 - 8 | 1079 | 69 | |
BC006156.1 | - | 6 - 7 | 795 | 69 | |
BC008325.1 | - | 6 - 7 | 1009 | 69 | |
BC038961.1 | - | 7 - 8 | 961 | 69 | |
BC065235.1 | - | 7 - 8 | 929 | 69 |