Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC45A1
Entrez Gene ID
50651
Gene Name
solute carrier family 45 member 1
Gene Aliases
DNB5
Chromosome Location
Chr.1: 8318085 - 8344167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080397.2 NP_001073866.2 3 - 4 617 71
XM_011541530.1 XP_011539832.1 3 - 4 594 71

View Full Product Details