The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080397.2 | NP_001073866.2 | 4 - 5 | 840 | 65 |
XM_011541530.1 | XP_011539832.1 | 4 - 5 | 817 | 65 | |
XM_011541531.1 | XP_011539833.1 | 3 - 4 | 724 | 65 |