Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGAP12
Entrez Gene ID
94134
Gene Name
Rho GTPase activating protein 12
Gene Aliases
-
Chromosome Location
Chr.10: 31805398 - 31928876 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270695.1 NP_001257624.1 14 - 15 2242 90
NM_001270696.1 NP_001257625.1 13 - 14 2167 90
NM_001270697.1 NP_001257626.1 14 - 15 2116 90
NM_001270698.1 NP_001257627.1 12 - 13 2026 90
NM_001270699.1 NP_001257628.1 11 - 12 1868 90
NM_018287.6 NP_060757.4 15 - 16 2257 90
XM_005252644.1 XP_005252701.1 13 - 14 2101 90
XM_011519761.1 XP_011518063.1 13 - 14 2035 90
XM_017016954.1 XP_016872443.1 11 - 12 1945 90
GenBank mRNA AK000744.1 - 11 - 12 1310 90
AK025438.1 - 8 - 9 667 90
AK091182.1 - 13 - 14 2112 90
AK315066.1 - 12 - 13 1919 90
AL137485.1 - 6 - 7 695 90
AL834250.2 - 11 - 12 1792 90
AY033594.1 - 13 - 14 2134 90
AY033595.1 - 15 - 16 2224 90
BC051811.1 - 3 - 4 279 90
BC094719.1 - 14 - 15 2166 90
BC115362.2 - 14 - 15 2013 90
BC115363.2 - 14 - 15 2013 90
CR936681.1 - 15 - 16 2143 90

View Full Product Details