The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130702.2 | NP_001124174.2 | 6 - 7 | 891 | 81 |
GenBank mRNA | AK027524.1 | - | 6 - 7 | 891 | 81 |