The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173216.2 | NP_775323.1 | 2 - 3 | 451 | 98 |
NM_173217.2 | NP_775324.1 | 2 - 3 | 451 | 98 | |
XM_006713734.1 | XP_006713797.1 | 3 - 4 | 706 | 98 | |
XM_011513085.1 | XP_011511387.1 | 3 - 4 | 714 | 98 | |
XM_017007065.1 | XP_016862554.1 | 4 - 5 | 798 | 98 | |
GenBank mRNA | A17362.1 | - | 2 - 3 | 256 | 98 |
DB235666.1 | - | 2 - 3 | 240 | 98 | |
X17247.1 | - | 2 - 3 | 250 | 98 |