Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
Chromosome Location
Chr.10: 97572441 - 97583884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129981.2 NP_001123453.1 6 - 7 1007 67
NM_001291219.1 NP_001278148.1 6 - 7 1007 67
GenBank mRNA BC020817.1 - 6 - 7 669 67

View Full Product Details