The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133494.2 | NP_598001.1 | 9 - 10 | 1131 | 73 |
XM_017000344.1 | XP_016855833.1 | 9 - 10 | 1133 | 73 | |
XM_017000345.1 | XP_016855834.1 | 10 - 11 | 1240 | 73 | |
GenBank mRNA | AB062450.1 | - | 8 - 9 | 850 | 73 |