The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000944.4 | NP_000935.1 | 9 - 10 | 1754 | 149 |
NM_001130691.1 | NP_001124163.1 | 9 - 10 | 1754 | 149 | |
XM_017008365.1 | XP_016863854.1 | 9 - 10 | 1058 | 149 | |
GenBank mRNA | AB451338.1 | - | 9 - 10 | 1079 | 149 |
AB451487.1 | - | 9 - 10 | 1079 | 149 | |
AK290532.1 | - | 9 - 10 | 1616 | 149 | |
AL353950.1 | - | 9 - 10 | 1485 | 149 | |
AY904364.1 | - | 8 - 9 | 924 | 149 | |
BC025714.1 | - | 9 - 10 | 1188 | 149 | |
EU192653.1 | - | 3 - 4 | 1071 | 149 | |
KU178269.1 | - | 9 - 10 | 1079 | 149 | |
KU178270.1 | - | 9 - 10 | 1079 | 149 | |
L14778.1 | - | 9 - 10 | 1226 | 149 |