The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013276.2 | NP_037408.2 | 2 - 3 | 412 | 100 |
GenBank mRNA | AF163573.1 | - | 2 - 3 | 412 | 100 |
AK055609.1 | - | 2 - 3 | 338 | 100 | |
AK312428.1 | - | 2 - 3 | 359 | 100 | |
AL832420.1 | - | 2 - 3 | 351 | 100 | |
BC020543.1 | - | 2 - 3 | 368 | 100 |