The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152573.3 | NP_689786.2 | 2 - 3 | 837 | 78 |
XM_005251730.3 | XP_005251787.1 | 2 - 3 | 886 | 78 | |
XM_005251731.3 | XP_005251788.1 | 2 - 3 | 1328 | 78 | |
GenBank mRNA | AK290986.1 | - | 2 - 3 | 579 | 78 |
BC023566.1 | - | 2 - 3 | 837 | 78 |